Please login to your online account to display your discounted pricing

Mouse anti-Macrophage Marker, Clone: 25F9, Biotin, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen Macrophage Marker
Clone 25F9
Immunogen The immunogen for this antibody is mature macrophage marker (late-stage).
Gene ID (Entrez) 9332
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP235326B1

 
novus biologicals canada
NBP235326B100UG
100μg Each for $362.00

NBP235326B3

 
novus biologicals canada
NBP235326B25UG
25μg Each for $175.00
Description & Specifications

Specifications

Antigen Macrophage Marker
Clone 25F9
Immunogen The immunogen for this antibody is mature macrophage marker (late-stage).
Gene ID (Entrez) 9332
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry