Please login to your online account to display your discounted pricing

Mouse anti-Ly-49G, Clone: AT-8, Biotin, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen Ly-49G
Clone AT-8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is Ly-49G (Ly49G).
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20035855

 
novus biologicals canada
NBP200358500UG
500μg Each for $329.00

NBP20035810

 
novus biologicals canada
NBP200358100UG
100μg Each for $131.00
Description & Specifications

Specifications

Antigen Ly-49G
Clone AT-8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is Ly-49G (Ly49G).
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry