Please login to your online account to display your discounted pricing

Mouse anti-Lung Specific Antigen, Clone: LSA120, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Lung Specific Antigen
Clone LSA120
Concentration 1mg/mL
Immunogen Protein extract of human lung cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23467701

 
novus biologicals canada
NBP23467701MG
0.1mg Each for $384.00

NBP23467702

 
novus biologicals canada
NBP23467702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Lung Specific Antigen
Clone LSA120
Concentration 1mg/mL
Immunogen Protein extract of human lung cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)