Please login to your online account to display your discounted pricing

Mouse anti-Lung Specific Antigen, Clone: LSA120, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467702

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Immunogen Protein extract of human lung cells
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone LSA120
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Host Species Mouse
Concentration 1mg/mL
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Antigen Lung Specific Antigen
Quantity 0.2mg
Regulatory Status RUO
Purification Method Purified
Conjugate Unlabeled