Please login to your online account to display your discounted pricing

Mouse anti-Lung Specific Antigen, Clone: LSA120, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467701

  • / Each
 

Description & Specifications

Specifications

Immunogen Protein extract of human lung cells
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen Lung Specific Antigen
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone LSA120
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Purification Method Purified
Primary or Secondary Primary
Concentration 1mg/mL