Please login to your online account to display your discounted pricing

Mouse anti-Luciferase (firefly), Clone: Luci17, Novus Biologicals™

$329.00 - $329.00

Specifications

Antigen Luciferase (firefly)
Clone Luci17
Concentration 1.0mg/mL
Immunogen Luciferase protein isolated from Photinus pyralis.
Formulation PBS. IgG purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17899705

 
novus biologicals canada
NBP178997005MG
0.05mg Each for $329.00
Description & Specifications

Specifications

Antigen Luciferase (firefly)
Clone Luci17
Concentration 1.0mg/mL
Immunogen Luciferase protein isolated from Photinus pyralis.
Formulation PBS. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Non-species specific
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting