Please login to your online account to display your discounted pricing

Mouse anti-Luciferase (firefly), Clone: Luci17, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178997005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17899705

  • / Each
 

Description & Specifications

Specifications

Format Purified
Monoclonal or Polyclonal Monoclonal
Antigen Luciferase (firefly)
Formulation PBS. IgG purified.
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Immunogen Luciferase protein isolated from Photinus pyralis.
Isotype IgG1
Purification Method Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity Detects Luciferase in C. elegans and Drosophilia melanogaster tissues, human fibroblast, mouse macrophage, kidney, liver and cortex as well as NIH3T3, Jurkat and BHR21 cell lines with little to no background signal.
Clone Luci17
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Non-species specific