Please login to your online account to display your discounted pricing

Mouse anti-LMO2, Clone: 1A9-3D11, Novus Biologicals™

$397.00 - $698.00

Specifications

Antigen LMO2
Clone 1A9-3D11
Immunogen Synthetic peptide corresponding to human LMO2 protein
Gene ID (Entrez) 4005
Formulation Prediluted Antibody. Tissue culture supernatant.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13606410

 
novus biologicals canada
NBP13606410ML
1mL Each for $698.00

NBP13606470

 
novus biologicals canada
NBP13606470ML
7mL Each for $397.00
Description & Specifications

Specifications

Antigen LMO2
Clone 1A9-3D11
Immunogen Synthetic peptide corresponding to human LMO2 protein
Gene ID (Entrez) 4005
Formulation Prediluted Antibody. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)