Please login to your online account to display your discounted pricing

Mouse anti-LMO2, Clone: 1A9-3D11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13606470ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13606470

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Format Supernatant
Isotype IgG1
Quantity 7mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Antigen LMO2
Formulation Prediluted Antibody. Tissue culture supernatant.
Host Species Mouse
Immunogen Synthetic peptide corresponding to human LMO2 protein
Purification Method Supernatant
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 4005
Test Specificity This product is specific for LMO2
Clone 1A9-3D11