Please login to your online account to display your discounted pricing

Mouse anti-LMO2, Clone: 1A9-3D11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13606410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13606410

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen LMO2
Conjugate Unlabeled
Host Species Mouse
Gene ID (Entrez) 4005
Immunogen Synthetic peptide corresponding to human LMO2 protein
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Format Supernatant
Formulation Prediluted Antibody. Tissue culture supernatant.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Quantity 1mL
Primary or Secondary Primary
Clone 1A9-3D11
Test Specificity This product is specific for LMO2
Purification Method Supernatant
Cross Reactivity Human