Please login to your online account to display your discounted pricing

Mouse anti-LIGHT/TNFSF14, Clone: 7-3 (7), APC, Novus Biologicals™

$175.00 - $395.00

Specifications

Antigen LIGHT/TNFSF14
Clone 7-3 (7)
Immunogen The immunogen for this antibody is TNFSF14.
Gene ID (Entrez) 8740
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20022625

 
novus biologicals canada
NBP20022625TESTS
25 Tests Each for $175.00

NBP200226

 
novus biologicals canada
NBP200226100TESTS
100 Tests Each for $395.00
Description & Specifications

Specifications

Antigen LIGHT/TNFSF14
Clone 7-3 (7)
Immunogen The immunogen for this antibody is TNFSF14.
Gene ID (Entrez) 8740
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry