Please login to your online account to display your discounted pricing

Mouse anti-LAT, Clone: LAT.10-17, APC, Novus Biologicals™

$175.00 - $395.00

Specifications

Antigen LAT
Clone LAT.10-17
Immunogen The immunogen for this antibody is LAT.
Gene ID (Entrez) 27040
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20029025

 
novus biologicals canada
NBP20029025TESTS
25 Tests Each for $175.00

NBP200290

 
novus biologicals canada
NBP200290100TESTS
100 Tests Each for $395.00
Description & Specifications

Specifications

Antigen LAT
Clone LAT.10-17
Immunogen The immunogen for this antibody is LAT.
Gene ID (Entrez) 27040
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Allophycocyanin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry