Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: SPM559, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Lambda light chain
Clone SPM559
Concentration 0.2mg/mL
Immunogen Purified human IgG
Gene ID (Entrez) 3536
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23437701

 
novus biologicals canada
NBP23437701ML
0.1mL Each for $139.00

NBP23437705

 
novus biologicals canada
NBP23437705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Lambda light chain
Clone SPM559
Concentration 0.2mg/mL
Immunogen Purified human IgG
Gene ID (Entrez) 3536
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting