Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: SPM559, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437705

  • / Each
 

Description & Specifications

Specifications

Antigen Lambda light chain
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3536
Clone SPM559
Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human IgG
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30.
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled