Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: SPM559, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442401

  • / Each
 

Description & Specifications

Specifications

Antigen Lambda light chain
Host Species Mouse
Immunogen Purified human IgG
Quantity 0.1mg
Gene ID (Entrez) 3536
Test Specificity This MAb is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Clone SPM559
Regulatory Status RUO
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Format Purified
Isotype IgG2a, κ
Primary or Secondary Primary
Concentration 1mg/mL
Conjugate Unlabeled