Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LcN-2 + ICO-106, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Lambda light chain
Clone LcN-2 + ICO-106
Concentration 0.2mg/mL
Immunogen Purified human IgG (LcN-2 and ICO-106)
Gene Accession No. P01701
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423601

 
novus biologicals canada
NBP23423601ML
0.1mL Each for $139.00

NBP23423605

 
novus biologicals canada
NBP23423605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Lambda light chain
Clone LcN-2 + ICO-106
Concentration 0.2mg/mL
Immunogen Purified human IgG (LcN-2 and ICO-106)
Gene Accession No. P01701
Gene ID (Entrez) 3536
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting