Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LcN-2 + ICO-106, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23423601ML

 View all options for this product


For Research Use Only

Catalog No. NBP23423601


Description & Specifications


Antigen Lambda light chain
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone LcN-2 + ICO-106
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01701
Host Species Mouse
Immunogen Purified human IgG (LcN-2 and ICO-106)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3536
Reconstitution Protein A purified