Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LcN-2 + ICO-106, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423605

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Purified human IgG (LcN-2 and ICO-106)
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Conjugate Unlabeled
Antigen Lambda light chain
Gene Accession No. P01701
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 3536
Reconstitution Protein A purified
Clone LcN-2 + ICO-106