missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LcN-2 + ICO-106, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Lambda light chain
Clone LcN-2 + ICO-106
Concentration 1mg/mL
Immunogen Purified human IgG (LcN-2 and ICO-106)
Gene Accession No. P01701
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23466501 Novus Biologicals Canada
NBP23466501MG
0.1mg Each for $384.00
NBP23466502 Novus Biologicals Canada
NBP23466502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Lambda light chain
1mg/mL
P01701
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
LcN-2 + ICO-106
Purified human IgG (LcN-2 and ICO-106)
3536
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot