Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LcN-2 + ICO-106, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466501

  • / Each
 

Description & Specifications

Specifications

Antigen Lambda light chain
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human IgG (LcN-2 and ICO-106)
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3536
Reconstitution Protein A purified
Clone LcN-2 + ICO-106
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P01701
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.