Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LcN-2 + ICO-106, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466502

  • / Each
 

Description & Specifications

Specifications

Antigen Lambda light chain
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone LcN-2 + ICO-106
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01701
Host Species Murine
Immunogen Purified human IgG (LcN-2 and ICO-106)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3536
Reconstitution Protein A purified
Target Species Human