Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LAM03, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423505

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01701
Host Species Mouse
Immunogen Purified human lambda light chain
Isotype IgG1, κ
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Antigen Lambda light chain
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 3536
Clone LAM03
Concentration 0.2mg/mL