Please login to your online account to display your discounted pricing

Mouse anti-Lambda light chain, Clone: LAM03, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen Purified human lambda light chain
Conjugate Unlabeled
Cross Reactivity Human
Antigen Lambda light chain
Formulation PBS with 0.05% BSA.
Gene Accession No. P01701
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3536
Reconstitution Protein A purified
Concentration 0.2mg/mL
Isotype IgG1, κ
Clone LAM03