Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: UJ127, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen L1CAM
Clone UJ127
Concentration 0.2mg/mL
Immunogen Homogenous suspension of 16 week human fetal brain
Gene Accession No. P32004
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23427305

 
novus biologicals canada
NBP23427305ML
0.5mL Each for $321.00

NBP23427301

 
novus biologicals canada
NBP23427301ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen L1CAM
Clone UJ127
Concentration 0.2mg/mL
Immunogen Homogenous suspension of 16 week human fetal brain
Gene Accession No. P32004
Gene ID (Entrez) 3897
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting