Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: UJ127, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Clone UJ127
Antigen L1CAM
Format Purified
Immunogen Homogenous suspension of 16 week human fetal brain
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 3897
Conjugate Unlabeled
Gene Accession No. P32004
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Reconstitution Protein A purified
Concentration 0.2mg/mL