Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: UJ127, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P32004
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Homogenous suspension of 16 week human fetal brain
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3897
Clone UJ127
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen L1CAM
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Reconstitution Protein A purified