Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: SPM275, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23301610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23301610

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen L1CAM
Format Purified
Gene Accession No. P32004
Host Species Mouse
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3897
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Immunogen Homogenous suspension of 16 week human fetal brain
Isotype IgG1, κ
Reconstitution Protein A purified
Clone SPM275
Conjugate Unlabeled