Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: SPM275, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23301601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23301601

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 3897
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Host Species Mouse
Quantity 0.1mL
Concentration 0.2mg/mL
Antigen L1CAM
Conjugate Unlabeled
Immunogen Homogenous suspension of 16 week human fetal brain
Formulation PBS with 0.05% BSA.
Gene Accession No. P32004
Reconstitution Protein A purified
Purification Method Purified
Clone SPM275