Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: SPM275, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475302

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Antigen L1CAM
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG1, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM275
Concentration 1mg/mL
Cross Reactivity Human
Gene Accession No. P32004
Immunogen Homogenous suspension of 16 week human fetal brain
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3897
Test Specificity Recognizes a cell surface protein of 220-240kDa, identified as L1 cell adhesion molecule.
Reconstitution Protein A purified
Conjugate Unlabeled