Please login to your online account to display your discounted pricing

Mouse anti-L1CAM, Clone: SPM275, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Conjugate Unlabeled
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunogen Homogenous suspension of 16 week human fetal brain
Gene Accession No. P32004
Isotype IgG1, κ
Format Purified
Host Species Mouse
Quantity 0.1mg
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Test Specificity Recognizes a cell surface protein of 220-240kDa, identified as L1 cell adhesion molecule.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Antigen L1CAM
Gene ID (Entrez) 3897
Clone SPM275
Reconstitution Protein A purified
Purification Method Purified