Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143849100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP143849

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Quantity 100 Tests
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Conjugate PE
Monoclonal or Polyclonal Monoclonal
Immunogen The immunogen for this antibody was PE CD62L.
Cross Reactivity Human
Regulatory Status RUO
Primary or Secondary Primary
Antigen L-Selectin/CD62L
Purification Method Affinity Purified
Formulation Phosphate buffer pH 7.2
Isotype IgG1, κ
Gene ID (Entrez) 6402
Format Affinity Purified
Concentration 0.2mg/mL
Clone DREG-56