Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14384925TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14384925

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Applications Flow Cytometry
Immunogen The immunogen for this antibody was PE CD62L.
Isotype IgG1, κ
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Clone DREG-56
Concentration 0.2mg/mL
Conjugate PE
Cross Reactivity Human
Antigen L-Selectin/CD62L
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 6402