Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, PE-Cy7, Novus Biologicals™

$192.00 - $354.00

Specifications

Antigen L-Selectin/CD62L
Clone DREG-56
Immunogen The immunogen for this antibody was CD62L.
Gene ID (Entrez) 6402
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142954

 
novus biologicals canada
NBP142954100TESTS
100 Tests Each for $354.00

NBP14295425

 
novus biologicals canada
NBP14295425TESTS
25 Tests Each for $192.00
Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Clone DREG-56
Immunogen The immunogen for this antibody was CD62L.
Gene ID (Entrez) 6402
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry