Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, PE-Cy7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142954100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142954

  • / Each
 

Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Applications Flow Cytometry
Clone DREG-56
Conjugate PE-Cy7
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was CD62L.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6402