Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, PE-Cy5, Novus Biologicals™

$171.00 - $321.00

Specifications

Antigen L-Selectin/CD62L
Clone DREG-56
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD62L.
Gene ID (Entrez) 6402
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14308825

 
novus biologicals canada
NBP14308825TESTS
25 Tests Each for $171.00

NBP143088

 
novus biologicals canada
NBP143088100TESTS
100 Tests Each for $321.00
Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Clone DREG-56
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD62L.
Gene ID (Entrez) 6402
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry