Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, PE-Cy5, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP143088100TESTS

 View all options for this product


For Research Use Only

Catalog No. NBP143088


Description & Specifications


Antigen L-Selectin/CD62L
Applications Flow Cytometry
Clone DREG-56
Concentration 0.2mg/mL
Conjugate PE-Cy5
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was CD62L.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6402