Please login to your online account to display your discounted pricing

Mouse anti-L-Selectin/CD62L, Clone: DREG-56, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200130100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP200130

  • / Each
 

Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Format Affinity Purified
Applications Flow Cytometry
Immunogen The immunogen for this antibody is L-selectin.
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Human
Host Species Mouse
Isotype IgG1, κ
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6402
Clone DREG-56
Conjugate FITC
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal