Please login to your online account to display your discounted pricing

Mouse anti-Ku70/XRCC6, Clone: KU729, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Ku70/XRCC6
Clone KU729
Concentration 0.2mg/mL
Immunogen Nuclear extract of human HL-60 cells
Gene Accession No. P12956
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23424701

 
novus biologicals canada
NBP23424701ML
0.1mL Each for $139.00

NBP23424705

 
novus biologicals canada
NBP23424705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Ku70/XRCC6
Clone KU729
Concentration 0.2mg/mL
Immunogen Nuclear extract of human HL-60 cells
Gene Accession No. P12956
Gene ID (Entrez) 2547
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation