Please login to your online account to display your discounted pricing

Mouse anti-Ku70/XRCC6, Clone: KU729, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424701

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen Ku70/XRCC6
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Nuclear extract of human HL-60 cells
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 2547
Test Specificity Does not react to Chicken, Mouse or Rat.
Reconstitution Protein A purified
Clone KU729
Cross Reactivity Human
Cross Reactivity Primate
Gene Accession No. P12956
Isotype IgG1, κ
Concentration 0.2mg/mL