Please login to your online account to display your discounted pricing

Mouse anti-Ku70/XRCC6, Clone: KU729, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424705

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Immunogen Nuclear extract of human HL-60 cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Cross Reactivity Human
Cross Reactivity Primate
Antigen Ku70/XRCC6
Gene Accession No. P12956
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2547
Test Specificity Does not react to Chicken, Mouse or Rat.
Reconstitution Protein A purified
Clone KU729
Conjugate Unlabeled