Please login to your online account to display your discounted pricing

Mouse anti-Ku70/XRCC6, Clone: KU729, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466301

  • / Each
 

Description & Specifications

Specifications

Immunogen Nuclear extract of human HL-60 cells
Monoclonal or Polyclonal Monoclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Cross Reactivity Human
Cross Reactivity Primate
Gene ID (Entrez) 2547
Clone KU729
Primary or Secondary Primary
Host Species Mouse
Gene Accession No. P12956
Regulatory Status RUO
Antigen Ku70/XRCC6
Test Specificity Does not react to Chicken, Mouse or Rat.
Concentration 1mg/mL
Quantity 0.1mg
Purification Method Purified
Conjugate Unlabeled
Isotype IgG1, κ
Reconstitution Protein A purified
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.