Please login to your online account to display your discounted pricing

Mouse anti-KLRA1, Clone: A1, PE, Novus Biologicals™

$171.00 - $362.00

Specifications

Antigen KLRA1
Clone A1
Immunogen The immunogen for this antibody was Ly49A.
Gene ID (Entrez) 10748
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14375701

 
novus biologicals canada
NBP14375701MG
0.1mg Each for $362.00

NBP14375703

 
novus biologicals canada
NBP1437570025MG
0.025mg Each for $171.00
Description & Specifications

Specifications

Antigen KLRA1
Clone A1
Immunogen The immunogen for this antibody was Ly49A.
Gene ID (Entrez) 10748
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry