Please login to your online account to display your discounted pricing

Mouse anti-KLRA1, Clone: A1, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1437570025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14375703

  • / Each
 

Description & Specifications

Specifications

Antigen KLRA1
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Mouse
Applications Flow Cytometry
Immunogen The immunogen for this antibody was Ly49A.
Quantity 0.025mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10748
Conjugate PE
Isotype IgG2a
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone A1
Cross Reactivity Mouse