Please login to your online account to display your discounted pricing

Mouse anti-KIR2DL5/CD158f, Clone: UP-R1, PE, Novus Biologicals™

$175.00 - $395.00

Specifications

Antigen KIR2DL5/CD158f
Clone UP-R1
Immunogen The immunogen for this antibody is KIR2DL5A.
Gene ID (Entrez) 57292
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200190

 
novus biologicals canada
NBP200190100TESTS
100 Tests Each for $395.00

NBP20019025

 
novus biologicals canada
NBP20019025TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen KIR2DL5/CD158f
Clone UP-R1
Immunogen The immunogen for this antibody is KIR2DL5A.
Gene ID (Entrez) 57292
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence