Please login to your online account to display your discounted pricing

Mouse anti-KIR2DL1/CD158a, Clone: HP-MA4, Cy5.5, PerCP, Novus Biologicals™

$197.00 - $428.00

Specifications

Antigen KIR2DL1/CD158a
Clone HP-MA4
Immunogen The immunogen for this antibody was CD158a.
Gene ID (Entrez) 3802
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142846

 
novus biologicals canada
NBP142846100TESTS
100 Tests Each for $428.00

NBP14284625

 
novus biologicals canada
NBP14284625TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen KIR2DL1/CD158a
Clone HP-MA4
Immunogen The immunogen for this antibody was CD158a.
Gene ID (Entrez) 3802
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b
Conjugate Cy5.5, PerCP
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry