Please login to your online account to display your discounted pricing

Mouse anti-KDEL, Clone: 10C3, Novus Biologicals™

$362.00 - $536.00

Specifications

Antigen KDEL
Clone 10C3
Immunogen Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78.
Gene ID (Entrez) 79070
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19746902

 
novus biologicals canada
NBP19746902MG
0.2mg Each for $536.00

NBP19746905

 
novus biologicals canada
NBP197469005MG
0.05mg Each for $362.00
Description & Specifications

Specifications

Antigen KDEL
Clone 10C3
Immunogen Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78.
Gene ID (Entrez) 79070
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Avian
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Invertebrate
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Plant
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Electron Microscopy
Applications Enzyme Immunoassay
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting