Please login to your online account to display your discounted pricing

Mouse anti-KDEL, Clone: 10C3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197469005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19746905

  • / Each
 

Description & Specifications

Specifications

Applications Electron Microscopy
Applications Enzyme Immunoassay
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Host Species Mouse
Conjugate Unlabeled
Antigen KDEL
Cross Reactivity Avian
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Invertebrate
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Plant
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Format Purified
Immunogen Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 79070
Purification Method Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Quantity 0.05mg
Isotype IgG2a
Test Specificity Recognizes proteins containing the KDEL sequence in mammals, primates, birds and Insects, Plants, and Yeast.
Clone 10C3
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.