Please login to your online account to display your discounted pricing

Mouse anti-KDEL, Clone: 10C3, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP19746902MG

 View all options for this product


For Research Use Only

Catalog No. NBP19746902


Description & Specifications


Antigen KDEL
Applications Electron Microscopy
Applications Enzyme Immunoassay
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 10C3
Conjugate Unlabeled
Cross Reactivity Avian
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Invertebrate
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Plant
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Yeast
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Host Species Mouse
Immunogen Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78.
Isotype IgG2a
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 79070
Test Specificity Recognizes proteins containing the KDEL sequence in mammals, primates, birds and Insects, Plants, and Yeast.