Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: TB28-2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23472101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23472101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Antigen Kappa Light Chain
Format Purified
Host Species Mouse
Immunogen Human IgG-kappa myeloma protein
Isotype IgG1, κ
Gene ID (Entrez) 3514
Clone TB28-2
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01601
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 1mg/mL
Cross Reactivity Human