Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: SPM558, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Kappa Light Chain
Clone SPM558
Concentration 0.2mg/mL
Immunogen Recombinant human Ig kappa chain
Gene ID (Entrez) 3514
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23437605

 
novus biologicals canada
NBP23437605ML
0.5mL Each for $321.00

NBP23437601

 
novus biologicals canada
NBP23437601ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen Kappa Light Chain
Clone SPM558
Concentration 0.2mg/mL
Immunogen Recombinant human Ig kappa chain
Gene ID (Entrez) 3514
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting